hotspots file
A BED or a VCF file that defines regions in the gene that typically contain variants and enables Ion Reporter™ Software to identify if a specific variant is present or absent. See also target regions file.