proband
A person or a sample that is serving as a starting point for the genetic study. Denoting the proband aids in establishing relationships within a group. In medical genetics, the proband is the first affected family member who seeks medical attention for a genetic disorder.