JavaScript must be enabled in order to use this site.Please enable JavaScript in your browser and refresh the page. base substitution classes Somatic mutations can be divided into six base substitution classes: C>A, C>G, C>T, T>A, T>C, and T>G. ×