Quality Control (QC) statistics files report coverage distribution. The QC files that contains
the statistics can be downloaded. QC report settings can be customized when you
create a custom analysis workflow or edit an analysis workflow.
QC package coverage distribution is calculated for each region and reported in the
No,
Low,
Medium, and
High columns in
the *_coverage_stastics.txt files.