Enable or disable private folders on Ion Reporter™ Server
An administrator can enable or disable a private folder in Ion Reporter™ Software for individual user accounts. Private folders allow a user to restrict all visibility to their data.
After a private folder is enabled, the data, analysis, and analysis workflows created by that user and generated in that folder are visible only to the user who owns the private folder, and to the administrator who set up the private folder.
This functionality is available only for the Ion Reporter™ Server. It is not available for Ion Reporter™ Software on Thermo Fisher™ Connect Platform.
×