The Ion Reporter™ Software web services API (application programming interface) can be used to automate returns and retrieve key information from the system. Ion Reporter™ Software APIs are compliant with REST (Representational State Transfer) architectural constraints.