The average number of reads representing a given nucleotide in the reconstructed sequence. Enables you to estimate the percentage of the genome covered by reads. High coverage overcomes errors in base-calling and assembly. The typical desired coverage of a genome is 30x.