amplicon coverage
An amplicon is a piece of DNA or RNA that is the source or product of a natural or artificial amplification or replication event. Coverage refers to the number of times the amplicon is amplified or replicated.