tumor mutational burden
A calculation of nonsynonomous variants (missense and nonsense single nucleotide variants (SNVs)) plus insertion and deletion variants (INDELs) detected per megabase (Mb) of exonic sequence.