Apply a sequence variant baseline workflow preset to an analysis workflow
You can create a custom Sequence Variant Baseline workflow preset that you can use as a baseline control to detect somatic variants. For more information, see Sequence variant baselines. You must add the custom baseline to an analysis workflow to use the baseline in an analysis.
- In the Workflows tab, start to create or copy an analysis workflow.
- In the Reference workflow bar step, select the target regions file for the panel that is associated with the analysis workflow.
- In the Parameters workflow bar step, click the link to open the Variant Finding parameters and click Advanced.
-
Scroll to select a sequence variant baseline
that matches the panel associated with the analysis workflow from the
Analysis SVB File
dropdown list.
After you make a selection, you can view details for the baseline in the Summary pane.
- Click Next.
-
Proceed through the remaining
steps in the workflow bar.
For more information, see Steps in analysis workflow creation.
- In the Confirm step, click Save Workflow.
×