You can view the analysis notes that are included in Ion Reporter™ Software. Analysis notes are time-stamped written notes to capture comments or observations for an analysis that has successfully completed. An analysis can contain multiple notes from different users.
You cannot edit analysis notes. However, you can add additional notes to an analysis. You can also add a description to an analysis. For more information, see Launch an analysis. Analyses notes can be shared within the same organization. Analysis notes are not shared when an analysis is shared with another Ion Reporter™ Software organization.
In the Analyses tab, select Overview.
To review analysis notes, on the Analyses Overview screen:
Click (Note) in the row for an analysis of interest.
Select the row of an analysis of interest, and then hover the mouse over the Notes: link in the Details pane of a selected analysis.