alpha-beta diversity
Alpha diversity results describe the diversity in a single sample at the Species, Genus, and Family levels. Beta diversity results describe the diversity between multiple samples at the Species, Genus, and Family levels. Used with Metagenomics 16S analysis workflows in Ion Reporter™ Software. See also metagenomics.