JavaScript must be enabled in order to use this site.Please enable JavaScript in your browser and refresh the page. Sort the Workflows table You can sort the Workflows table to make it easier to find the analysis workflows that you are looking for in Ion Reporter™ Software. In the Workflows tab, in the Overview screen, click a column heading to sort the list of analysis workflows. Click the column again to reverse the order. Click the column a third time to return the column to the order that was used before the sort and stop sorting for the column. Related Topics Parent topic: Find analysis workflows ×