You can lock an analysis workflow to ensure that the settings are not changed. After an analysis workflow is locked, it cannot be edited or unlocked. However, it can be copied.
Click the Workflows tab.
Select the analysis workflow that you want to lock, then click Actions > Lock.
Click Yes to confirm that you want to lock the analysis workflow.
Due to a database update latency, it can take a few minutes for the analysis
workflow to lock.
When the workflow is locked, a (Lock) is shown
beside the name of the workflow in the list of analysis workflows. The analysis
workflow is now locked and cannot be edited.