Ion Mesh
A network of Ion Torrent™ Servers that allows users to perform the following actions:
  • View all runs of interest across multiple servers on the same data page.
  • Transfer Planned Runs between different connected servers.
  • Perform flexible workflows for Ion 550™ Chips across different Ion Chef™ instruments connected to different servers. Torrent Suite™ Software can track reagent/cartridge usage across multiple servers that are a part of the same Ion Mesh.