JavaScript must be enabled in order to use this site.Please enable JavaScript in your browser and refresh the page. functional score A filter in Ion Reporter™ Software that provides functional scores based on SIFT, PolyPhen, and Grantham scores. See also SIFT score, PolyPhen score, and Grantham score. ×