Visualization of genomic segmentation analysis, Allele Specific Copy Number plots
- Do one of the following to visualize analysis results that used an analysis workflow to detects genomic segmentation and allele specific copy number.
- Click the Allele Specific Copy Number tab.
A series of plots showing copy number variation across the entire genome is displayed. Click Export Image to save a PNG image of all three plots to a location of your choice. If visualizing multiple analyses simultaneously, individual sample results are listed sequentially.
This visualization shows identified genomic segments (shown as horizontal lines) using log2 ratios from the Copy Number Variation (CNV) analysis and log odds from the Torrent Variant Caller (TVC) analysis. Segments are identified using the heterozygous population SNPs (shown as dots in the upper two panels) targeted by assay amplicons.
Plot scale (x-axis)
Description
logR = copy number log2 ratio
Log2 ratios (top panel) of the copy-number estimates relative to the baseline copy-number as calculated by the CNV algorithm for each amplicon in the assay. Ratio representative Genome segmentation overlay (horizontal brown bars) shows segments with similar log2 ratios clustered together.
log odds = ln(Tumor Reads / Reference Reads)
Log odds (middle panel) for each heterozygous SNP in the assay. Log odds is calculated as the natural logarithm of the ratio of the sequencing reads with variant allele and reference allele that are obtained using Torrent Variant Caller (TVC). Genome segmentation overlay (horizontal brown bars) shows segments with similar log odds clustered together. Since the variant allele could be major or minor for any SNP, the corresponding log odds could be positive or negative and are therfore displayed as segments that are mirror images around 0.
Tumor Copy Number
Bottom panel shows the total (black horizontal line) and minor (red horizontal line) copy-number estimates for each of the identified genomic segments (top two panels). Genomic segments where total CN ≥1 and minor CN = 0 are segments with LOH. There may exist genomic segments for which minor copy-number estimates can not be determined.