CFTR analysis results
Review the following examples for information about how to review data in the analyses results from CFTR workflows:
  • The Variant Name column reports matches with the Named Variants annotation source.
  • The following IGV visualization for CFTR analysis results include tracks for long deletions:
  • The following IGV visualization for CFTR analysis results include a named variant:
  • The following IGV visualization for CFTR analysis results include a hotspot position:
  • Hover the mouse over a position in the IGV visualization to view details such as the following:
The factory-provided CFTR workflow includes long deletion detection. You can copy/edit the CFTR workflow and preserve long deletion detection. If you create your own CFTR workflow (even with the CFTR regions file), your workflow includes the standard Ion AmpliSeq workflow template and does not include the long deletion detection module.