An administrator can track system activities occurring within the organization in Ion Reporter™ Software. For example, you can see who created analyses and who modified samples. Using this feature, you can select a time period to audit.
This feature allows you to meet Title 21 CFR Part 11 of Federal Regulations that establishes the United States Food and Drug Administration regulations on electronic records and signatures, password policies, and user activity auditing.