Sample presets
You can create custom sample attributes, which are known as sample presets, to describe samples that are used in your research.
Ion Reporter™ Software supports the following types of attributes:
-
Boolean—Creates a list of these two choices: True, False.
-
Text—Accepted values: general text.
-
Integer—Accepted values: integer numbers. Examples: 0, 9, −44.
-
Float—Accepted values: decimal numbers Examples: 0.1, 9.0, −44.3.
-
Controlled Vocabulary—Creates a list of these choices that you define with the preset terms.
-
Date—Accepted values: provides a calendar tool to select a date.
Ion Reporter™ Software gives you the option to encrypt the internal storage for a sample attribute.
×