JavaScript must be enabled in order to use this site.Please enable JavaScript in your browser and refresh the page. Sort the Sample Presets table You can sort the Sample Presets table to organize the information by Sample Preset Name, User, or Date of Creation. In the Samples tab, click Presets. In the Sample Presets table, click the Name, Created By, or Created On column heading to sort the list. Click the column heading again to reverse the order. Related Topics Parent topic: Sample presets ×