Apply a preference to the Samples table
You can apply saved table preferences to the Samples table in Ion Reporter™ Software.
  1. In the Samples tab, click Overview.
  2. Click PreferencesTable Preferences.
    A menu of saved preferences appears.
  3. Select the preference from the menu.
The table preference is applied to the Samples table. The last saved table preference remains until another preference is selected or default settings are restored.