The Samples table is organized into columns of sample attributes that are initially organized in a default order. You can customize the table with table preferences to determine which columns appear and where the columns are placed. Use table preferences to include or exclude columns, then drag and drop each column to the desired position in the table. You can also use the column headings to resize the default width of each column.