Download a file
|
download |
|
|---|---|
|
URL |
/api/v1/download |
|
Description |
Gets the file, creates a compressed archive file in ZIP file format and downloads it. |
|
Method |
GET |
|
Request query parameters |
|
|
Example request |
|
|
Request headers |
Content-Type: application/x-www-form-urlencoded Authorization:API Token |
|
Returns |
Downloads the file that is located at the file path that is specified. |
Response fields
|
Status code |
Response message |
|---|---|
|
200 |
Successful response |
|
400 |
Bad request |
|
404 |
Not found |
|
401 |
User is not authenticated |
|
500 |
Error occurred |
Curl command
curl -O -v -k -X GET "https://<server name>/api/v1/download?filePath=/data/IR/data/IR_Org/data/IRU_Uploads/2016-7-26_23_8_42/v2/report.pdf" -H "Content-Type:application/x-www-form-urlencoded" -H "Authorization:NDliYmU0OWE4MzA3YmE2MWVmMjhlZTlhOTEyMmE5ODM0NzU3NjQ4OGM5NzVjODU3YjdlMWM4Mjk3OGYzODE4Yw"