This filter can be added to filter chains in the software to detect .
Filter conditions include the ability to include (In) or exclude (Not In) hotspot position overlaps in filtered analysis results.
The filter returns variants that overlap hotspots that are defined in an uploaded hotspots file.