This filter can be added to filter chains in the software to determine the p-value of variants. The p-value is a statistical method for the detection of variant calls from next-generation sequencers.
Filter conditions include the ability to set a range from 0.0 to 1.0. In addition, the range boundary values can be included or excluded, and unannotated variants can be included or excluded in filtered analysis results.
The filter returns p-values for variants calls in the samples.