-
Launch the analysis.
|
From the…
|
Directions
|
|
Home tab
|
-
In the Dashboard screen, click Launch analysis.
-
In the Launch Analysis screen, select a Research Application.
-
In the Research Category list, select a row with an analysis workflow name.
-
Click Next.
|
|
Analyses tab
|
-
Click Launch Analysis, then select Manual from the list.
-
In the Launch Analysis screen, select a Research Application.
-
Click in the analysis workflow row, then click Next.
|
|
Workflows tab
|
-
Select a Research Application.
-
Click in the analysis workflow row, then in the Details pane select Launch Analysis from the Actions list.
|
-
Search by any unique identifier you used to label your samples during setup. Select one or more of the samples to include in the analysis, then click Next.
-
(Optional) Select plugins to run with your analysis.
Note: The Oncomine™ Variant Annotator plugin is applied by default and cannot be edited or removed from the
analysis workflow.
-
Click
(Actions) adjacent to the selected plugin.
-
Configure the plugin parameters as necessary, click Submit, then click Close.
-
Click Next.
-
Enter an Analysis Name and an optional Description, then click Launch Analysis.
The Ion Reporter™ Software performs the analysis and the results appear on the Analyses page.