Search for samples
You can search the Samples table for all sample attributes that are shown in the Samples table at the time of the search. Both predefined and custom sample attributes are included in the search results.
IMPORTANT! Search terms must adhere to the following rules.
-
An asterisk (*) is not allowed in the search field for use as a wildcard.
-
Searches are case-insensitive, that is, both upper-case and lower-cases letters are found regardless the case of search term letters.
-
Searches find every occurrence of a continuous string. For example, a search for demo in analysis names returns a list of all analysis with a name that includes demo. For example, demo1, demo2, and so on.
-
Spaces are removed during searches, and therefore are not recommended. For example, a search for demo 1 would return results only for data that includes the string demo1.