Sample ID
|
Name of the sequenced sample imported from a sequencing run.
|
SNV/INDEL
|
Gene
|
HGNC reviewed official gene symbol.
|
AA Chg
|
Amino acid change resulting from non-synonymous DNA variant.
|
Mutant Frequency %
|
Frequency of mutant allele expressed as a percentage.
|
Oncomine Variant Class
|
Variant class annotation that provides a high-level summary of the variant type (hotspot, fusion, amplification, and so on). The annotation is included when the Oncomine™ Variant Annotator plugin is run in Ion Reporter™ Software.
|
Oncomine Gene Class
|
Variant gene functional annotation that provides a high-level summary of gene type (Gain-of-Function, Loss-of-Function, or Unclassified). The annotation is included when the Oncomine™ Variant Annotator plugin is run in Ion Reporter™ Software.
|
Info
|
HS (targeted hotspot) or PN (potentially novel TP53 variant). De novo variant calls available for the breast panel only.
|
Genotype
|
Genotype measured associated with a DNA variant call.
|
Ref Allele
|
Reference allele as defined in the human genome reference (hg19).
|
Mut Molecular Cov.
|
Molecular coverage of the mutant allele.
|
WT Molecular Cov.
|
Molecular coverage of the wild type allele from the reference genome.
|
Depth
|
Total read coverage across amplicon containing SNV/INDEL hotspot locations. Count of chip-level reads aligned at this locus that participate in variant calling.
Note: This description applies to both, Ion AmpliSeq™ HD and non‑Ion AmpliSeq™ HD analyses. |
QC Test (LOD) %
|
Quality control check for SNV/INDEL target regions based on molecular coverage.
|
Transcript ID
|
NCBI accession number for the transcript representing the gene target being measured.
|
Locus
|
Chromosome and position of detected variant. Click the hyperlink to open the Ion Reporter™ Genomic Viewer to the specified locus.
|
CNV
|
Gene
|
Gene locus targeted for CNV measurement.
|
Gain/Loss
|
Detected copy number gain or loss.
|
CNV ratio
|
Ratio of measured CNV gene locus coverage relative to coverage on non-CNV loci.
|
p-value
|
Significance of CNV ratio measurement.
|
Med. Mol Cov. Gene
|
Median molecular coverage of targeted CNV gene.
|
Med. Mol Cov. Ref
|
Median molecular coverage of non-CNV reference loci.
|
Med. Read Cov. Gene
|
Median read coverage of targeted CNV gene.
|
Med. Read Cov. Ref
|
Median read coverage of non-CNV reference loci.
|
QC Test
|
Assay quality control as determined by amplicon coverage uniformity and number of amplicons remaining after outlier removal.
|
Valid CNV Amplicons
|
Number of CNV amplicons remaining after outlier removal.
|
CNV Locus
|
Chromosomal location of CNV gene being targeted.
|
Fusion
|
Variant (exons)
|
Name of fusion targeted and respective acceptor and donor exons.
|
Oncomine Driver Gene
|
The gene thought to be associated with increased oncogenic properties due to inappropriate activation by the fusion.
|
COSMIC/NCBI
|
COSMIC mutation or NCBI accession number.
|
Mol Cov. Mutant
|
Median molecular coverage across fusion amplicon.
|
Read Cov. Mutant
|
Median read coverage across fusion amplicon.
|
Detection
|
Detection status from assay.
|
QC Test
|
Assay quality control measured from expression detection of housekeeping genes.
|
Type
|
Assay type (For example, Fusion, RNA exon variant (exon skipping), Proc Control).
|
Locus
|
Chromosomal locations of targets included in assay.
|
Ratio To Wild Type
|
Ratio molecular for exon skipping assay relative to wild type control amplicons.
|
Norm count Within Gene
|
Exon skipping assay coverage normalized to molecular coverage of wild type (WT) MET control amplicons. (Lung panel only).
|