Sample definition
Before samples can be analyzed in Ion Reporter™ Software, samples must first be defined if any of the following is true:
  • Data are from BAM files that are uploaded manually to Ion Reporter™ Software from Torrent Suite™ Software, using the Upload Only option.
  • Data are imported manually through BAM or VCF files to Ion Reporter™ Software.
  • Data are contained in a CSV file of samples that was uploaded to Ion Reporter™ Software.
The sample definition process assigns a unique name to a collection of one or more sample files and the corresponding sample attributes. After samples are defined, sample data can be included when you launch an analysis in Ion Reporter™ Software.