Archive samples to an external storage device
This information applies only to Ion Reporter™ Server.
You can archive samples from an Ion Reporter™ Server to an external storage device to increase disk space.
Available options for external storage devices include the following.
  • You need to have an external storage device that is mounted to your Ion Reporter™ Server to archive samples to the device.
  • If you have a new Ion Reporter™ Server or upgrade from a previous version, you need to specify a storage location for archiving samples and analyses in Ion Reporter™ Server.
  • You can only archive samples if no processes, such as an analysis is running, or new sample uploads in progress, are active on the server.
  1. In the Samples tab, in the Overview screen, select the sample or samples to be archived.
  2. Complete one of the following actions.
    Option
    Action
    Archive multiple samples
    In the Selected Samples section, click ActionsArchive.
    Archive a single sample
    In the Details section, click ActionsArchive .
  3. Indicate whether the archives should be retained on the server or deleted in the Confirm Batch Archive dialog box.
    • To delete the samples after the archival process is complete, select the option do you want to delete the data from path post archive?.
    • To retain the samples after the archival process is complete, deselect the option do you want to delete the data from path post archive?.
  4. Click Yes in the Confirm Batch Archive dialog box to begin the archive process for the selected samples.
    The Archival Manager screen shows the progress of the archive.
  5. (Optional) To stop the archive process, click Abort.
    To prevent a loss of data, if you archive multiple analysis you can only abort the archive process if none of the archival processes are complete. For example, if there are 3 archives that are in progress, and 2 archives are completed, but the third item is in progress, the abort does not occur.
When the archival process is successful, there is a message in the Confirm Batch Archive dialog box to indicate that the process is complete. The samples are listed in the Archives screen. A successful archive has a Status of Restored. The table shows the name, type, size, status and any error message for the restoration.