Find samples
You can search, sort, and filter the list of samples in the Samples table in Ion Reporter™ Software.
Note: You can add attributes that are not shown in the Sample table, and later search for those attributes. For more information, see Customize the Samples table.
×