Visualize analysis results with Ion Reporter™ Software
You can visualize any human DNA or RNA analysis type with Ion Reporter™ Software for research use. You can also visualize multiple analyses in a single browser tab that is useful for comparisons.
Ion Reporter™ Genomic Viewer (IRGV) is a streamlined version of the Integrative Genomics Viewer (IGV) that is developed by the Broad Institute. IRGV is included with Ion Reporter™ Software. You can set preferences that specify which viewer opens by default and which elements are included in IRGV visualizations. For more information, see Set IRGV or IGV as the default viewer and Set IRGV preferences.