JavaScript must be enabled in order to use this site.Please enable JavaScript in your browser and refresh the page. Coverage histogram When you zoom in on the pileup, you see the coverage histogram and reads from the .bam track. The histogram in gray shows read depth at that location and reads from the .bam track show read direction and location of variants. Read depth Read location and direction of variants Related Topics Parent topic: Visualize analysis results with Ion Reporter™ Software ×