non-PAR
Non-Pseudoautosomal Regions (PAR 1 and PAR 2) of the human X and Y chromosomes pair and recombine during meiosis. Therefore, genes in this region are not inherited in a strictly sex-linked fashion.