Confirm the presence of fusion calls in Ion Reporter™ Software
Analysis results in Ion Reporter™ Software provide several ways to confirm a fusion call, including Imbalance values, visualization, and Normalized Detection Fractions. For more information, see Novel fusion detection using expression imbalance, View RNA Exon Tile Fusion Imbalance plots, and 3'/5' Imbalance scores for assay calls.
×