Filter chains for fusion panels
For Oncomine™ fusion panels, the Oncomine Variants filter chain is applied by default. There are many other filter chains for use with all other fusion panels and filters for non-Oncomine™ Fusion panels. You can use the Default Fusion View filter chain for other fusion panels, including custom panels from AmpliSeq.com.
-
For analysis results that include only Present fusion calls, click the Filter menu.
-
Select the Default Fusions View filter.
Note: The following data types along with present fusions calls will always be displayed as Filtered In variants in the Type column, whether the Default Fusions View filter is set to Present or Absent:
-
EXPR_CONTROL
-
ASSAYS_5P_3P
-
GENE_EXPRESSION