Filtered Coverage filter
This filter can be added to filter chains in the software to review results that meet a specific coverage threshold. By default, the filter reports all results with a coverage threshold greater than 100. Or, you can use the filter settings to change the threshold to include or exclude boundary values for the threshold, and to include or exclude unannotated variants in filtered analysis results. The default threshold setting is 100.
Filter conditions include the ability to set a minimum and a maximum value for minor allele frequency for the coverage threshold. Ranges from 0.0 to 0.0045 can be selected for the minimum in the From field and in the To field for the maximum. In addition, the range boundary values can be included or excluded, and unannotated variants can be included or excluded in filtered analysis results.