Amplicon coverage reports
Amplicon coverage reports list amplicons, SNPs, and bases in the amplicons. When you view the amplicon coverage report, you can adjust the amplicon coverage threshold to focus your report on only the amplicons that met the threshold that you set.
×