Software is unresponsive and cannot be rebooted
Software screen freezes or is unresponsive
When using a local Ion Reporter™ Server, the Ion Reporter™ Software screen freezes or becomes unresponsive, and you are unable to open the software, and/or unable to reboot the software.
Possible cause
Recommended action
Disk crash on Ion Reporter™ Server.
Work with your Field Support Engineer (FSE) or Technical Support to back up data.