Search for analysis workflows
You can use keywords or text strings to search for data in the Workflow Name column of the Workflows table.
IMPORTANT! Search terms must adhere to the following rules.
-
An asterisk (*) is not allowed in the search field for use as a wildcard.
-
Searches are case-insensitive, that is, both upper-case and lower-cases letters are found regardless the case of search term letters.
-
Searches find every occurrence of a continuous string. For example, a search for demo in analysis names returns a list of all analyses with a name that includes demo. For example, demo1, demo2, and so on.
-
Spaces are removed during searches, and therefore are not recommended. For example, a search for demo 1 would return results only for data that includes the string demo1.
- In the Workflows tab, click Overview.
- In the Search box, enter a search term, then click Go to return a list of workflows that match the keyword.
If the search string is invalid, the search field is outlined in red. Correct the search term to proceed.
The search results are returned in the Workflows table.