View analysis workflow details
When you select a single analysis workflow that is listed in the Workflows table in Ion Reporter™ Software, you can view information about its components in the Details section.
  1. Click the Workflows tab.
  2. On the Overview screen, in the Workflows table, select a row.
  3. In the Details section, view the analysis workflow details.
    The Details section is displayed.
    The information that is available in the Details section depends on how you defined the analysis workflow. For more information, see Analysis workflow details.
    Custom analysis workflows can be edited from the Details section. Predefined analysis workflows cannot be edited.
  4. In the Details section, click the View link to open a dialog box that displays the workflow parameters.
Analysis workflow details
When you select a single analysis workflow that is listed in the Workflows table in Ion Reporter™ Software, you can view information about its components in the Details section. The information that is available depends on the settings in the analysis workflow.
Custom analysis workflows can be edited from the Details section. Predefined analysis workflows cannot be edited. For more information, see Edit a user-defined analysis workflow.
Analysis workflow detail
Description
Research Application
The research application in use in the analysis workflow.
Workflow name
The name and version of the analysis workflow.
Description
A short description of the analysis workflow.
Version
The version number of Ion Reporter™ Software.
Revision
The revision number of the analysis workflow.
Sample Group
The sample group in use in the analysis workflow.
Research Category
The research category selected for the analysis workflow.
Reference
The reference selected for the analysis workflow.
Target Regions
The file name of the target regions file used in the analysis workflow.
Variant Type Detection
The variant type detection selected for the analysis workflow.
Hotspot Regions
The file name of the hotspots file used in the analysis workflow.
Fusion Panel
The name and version of the fusion panel.
MSI Baseline File
The file name of the MSI baseline file used in the analysis workflow. This file contains baseline information that accounts for variations in sample preparation or run conditions to support robust MSI detection.
MSI Marker Regions
The file name of the MSI marker regions file used in the analysis workflow. The MSI marker regions file contains information about the genomic position of the MSI markers, and more information that is used by the MSI detection algorithm.
TMB Filter Chain
The TMB Filter Chain that is used in the analysis workflow.
TMB Calculation Version
The version of the tumor mutational burden algorithm that is available in the analysis workflow is listed. The algorithm version can provide information about how the data was analyzed and can be useful if earlier versions of the software were used, or if troubleshooting is required.
Sequence Variant Baseline
The file name of the Sequence Variant Baseline file used in the analysis workflow.
Annotations
The annotations that are used in the analysis workflow, based on the annotation set that is selected.
MyVariants Database
The MyVariants database in use in the analysis workflow.
Filter Chain
The filter chain that is in use for the analysis workflow.
Copy Number
The copy number baseline control in use in the analysis workflow. The copy number baseline identifies the copy number variants (CNVs) in samples.
Plugins
The name and version number of the plugin or plugins in use in the analysis workflow.
Report Template
The name of the report template used in the analysis workflow.
Modified By
The Ion Reporter™ Software user who modified the analysis workflow.
Modified On
The date and time that the analysis workflow was modified.
Created By
The Ion Reporter™ Software user who created the analysis workflow.
Created On
The date and time that the analysis workflow was created.
Parameters
Click the View link to open a window to see the parameters that are defined for the selected analysis workflow.
Tag for IRU
Indicates whether the analysis workflow is tagged for use with IonReporterUploader plugin.