Analysis workflow revision autonumbering
Ion Reporter™ Software displays revision numbers of analysis workflows, and automatically increases the revision numbers when you copy or edit an analysis workflow.
To view analysis workflow version numbers, click WorkflowsOverview, then select the desired analysis workflow. The analysis workflow version number, for example w2.2, appears in two places.
  • In the Workflow Name column at the end of the analysis workflow name.
  • In the Details section at the end of the analysis workflow name.
The version number increases if you copy a predefined analysis workflow, or copy and edit a user-defined analysis workflow and use the name of the previous analysis workflow as the name for the new analysis workflow.
You can copy predefined analysis workflows and user-defined analysis workflows from the current version of the software or from previous versions of the software.