Main tab
|
Maximum Coverage
|
The maximum coverage of locations in the reference. Locations with coverage more than the maximum coverage values are ignored during coverage calculations.
Allowed values: 100 to 1,000,000
|
Maximum read length
|
The maximum read length.
Allowed values: 700 to 20,000
|
Advanced tab
|
Max mapping qv
|
The maximum mapping quality value. Any alignment with mapping quality value more than the specified value is ignored.
Allowed values: 0 to 255
|
Max mismatches
|
The maximum number of mismatches allowed in the alignments. Any alignment with more than the specified number of mismatches are ignored while generating reports related to the number of mismatches.
Allowed values: 0 to 500
|
Max base qv
|
The maximum base quality values. Any base with a base quality value of more than the specified value are ignored while generating reports.
Allowed values: 0 to 100
|
Maximum Medium Coverage (QC)
|
The maximum medium coverage value. Any base with a coverage value of more than the specified value are ignored while generating reports. The value of Maximum Medium Coverage (QC) should be greater than Maximum Low Coverage (QC).
Allowed values: 2 to unlimited
|
Minimum Target Overlap Forward
|
The fraction of an alignment that must be overlapped by a target in order to be classified as on target.
Allowed values: 0.000001 to 1
|
Maximum Low Coverage (QC)
|
The maximum low coverage value. Any base with a coverage value of more than the specified value is ignored while generating reports. Value of Maximum Low Coverage (QC) should be less than Maximum Medium Coverage (QC).
Allowed values: 1 to unlimited
|
Maximum Target Overlap Reverse
|
The fraction of an alignment that must be overlapped by a target in order to be classified as on target.
Allowed values: 0.0001 to 1
|