Download QC and final reports
To retrieve one Quality Control (QC) report in JSON format, enter:
curl --request GET -k -H "Authorization:ZDMxNDUwZGYyNjM4NGRlZjY0NjAyNTc1YTE1ZjU2MzFkNjVmYzQxYjJhOTQwNTYyYWM3OTJmZDY2YTU5NGQwNA" -O "https://<server name>/api/v1/download?filepath=/data/IR/data/IR_Org/ion.reporter@lifetech.com/new_CFTR/new_CFTR_20151106002127527/new_CFTR_1446798039543.zip"
The output in JSON format will resemble this:
[ { "reports": { "qc": { "link": "https://<server name>/api/v1/download?filepath=/data/IR/data/IR_Org/download/pdf/335a1f5e-19f1-4ff4-b80c-af620f1dfe1f/ff80818150dbc2430150dbe2d9650072_QC.pdf" }, "final": {} }, "report_published": "", "workflow": "AmpliSeq CFTR single sample", "data_links": { "filtered_variants": "https://<server name>/api/v1/download?filepath=/data/IR/data/IR_Org/ion.reporter@lifetech.com/new_CFTR/new_CFTR_20151106002127527/new_CFTR_1446798039543.zip", "unfiltered_variants": "https://<server name>/api/v1/download?filepath=/data/IR/data/IR_Org/ion.reporter@lifetech.com/new_CFTR/new_CFTR_20151106002127527/new_CFTR_1446798039543.zip" }, "variants_saved": "", "samples": { "PROBAND": "new_CFTR" }, "shared_with": [], "stage": "Review Variants", "flagged": false, "name": "new_CFTR_1446798039543", "id": "ff80818150dbc2430150dbe2d9650072", "ion_reporter_version": "5.0", "started_by": "Ion User", "start_date": "November 06, 2015", "status": "SUCCESSFUL" } ]
To retrieve all of the QC reports in JSON format, enter:
curl --request GET -k -H "Authorization:ODkxY2IxYThhZWNjNDU3MjlmNjdlZDBkYWEwYzdmZTQzODFmZjk3NjExNDA0ODVmYjYwMjA4YzQ5MGEwNDEzZg" https://<server name>/api/v1/qcreport?format=json
To retrieve one final report in JSON format, enter:
curl --request GET -k -H "Authorization:YTJjNGVmYWYyNDcxNTgyMmU3NzMxZmY0OWVjY2ZhYTA0ZDM2YTk1OTQ4ZjUxZDlmMzFkZjFjM2UwNzZhNjliMg" "https://<server name>/api/v1/finalreport?format=json&name=testDNAFusion_c255_1421470231458"
×