Download a BAM file
|
downloadBAM |
|
|---|---|
|
URL |
/api/v1/downloadBAM |
|
Description |
Download a BAM file for the sample or analysis in the filepath entered. This procedure applies to Ion Reporter™ Server only. |
|
Method |
GET |
|
Request query parameters |
|
|
Example request |
|
|
Request headers |
Content-Type: application/x-www-form-urlencoded Authorization:API Token |
|
Returns |
Downloads the BAM file in the filepath entered. User can replace "download.bam" in curl command with the BAM file name to change the name of the BAM file downloaded. |
Response fields
|
Status code |
Response message |
|---|---|
|
200 |
Successful response |
|
400 |
Bad request |
|
404 |
Not found |
|
401 |
User is not authenticated |
|
500 |
Error occurred |
Curl command
curl -o download.bam -L -k -X GET "https://<server name>/api/v1/downloadBAM?filePath=/data/IR/data/IR_Org/ion.reporter@lifetech.com/Demo_AmpliSeq_CHPv2_tumor/Demo_AmpliSeq_CHPv2_tumor_20180319063625805/outputs/VariantCallerActor-00/merged.bam.ptrim.bam" -H "Content-Type:application/x-www-form-urlencoded" -H "auth: ZWJjODU2ODIwYWY4OTk2Yjg2MzI5NTU1MzIxZTA0NGVlYTZlNjIyYTJlMGJlNDMwMGI3OWM4ZGNhZTNlNzg1Mw" -H "Connection: close"