You can import MyVariants from a VCF or a TSV file that is exported from another MyVariants database, or from a TSV file that you create. For more information, see Export MyVariants database.
In the Analyses tab, click MyVariants, then click Import MyVariants.
In the Upload MyVariants File screen, click Select file , then browse to the VCF or TSV file.
Click Submit.
When the import is complete, a message indicates that the file has been uploaded and lists the number of records that were uploaded, and the number of records that are valid and invalid.