JavaScript must be enabled in order to use this site.Please enable JavaScript in your browser and refresh the page. Delete an Analysis Results table preference You can delete a custom table preference setting for the Analysis Results table in Ion Reporter™ Software. In the Analyses tab, click Overview. Click the link for an analysis of interest to open the Analysis Results table. Click Preferences > Table Preferences, then select the custom table preference that you want to delete. Click Preferences > Delete Table Preferences. In the Confirm Delete dialog box, click Yes to confirm the deletion. The custom table preference is removed from the Table Preferences list. Related Topics Parent topic: Customize the Analysis Results table ×