JavaScript must be enabled in order to use this site.Please enable JavaScript in your browser and refresh the page. Restore default settings for the Analyses table You can restore default table column headings in the Analyses table in Ion Reporter™ Software. In the Analyses tab, click Overview. Click Preferences > Restore Defaults. The default table settings are restored. Related Topics Parent topic: Customize the Analyses table ×