Create an Analyses table preference
You can create a table preference setting in Ion Reporter™ Software. The table preference enables you to view only the columns of data that are relevant to your research in an Analyses table.
The user-defined table preference setting is included as output when you download a variants file.
  1. In the Analyses tab, click Overview.
  2. Click PreferencesSelect Columns, then select the sample attributes that you want to add to the Samples table, or deselect the sample attributes that you want to remove from the table.
    The selected columns are added on the right side of the table.
  3. Save the custom table as a preference.
    Option
    Description
    PreferencesSave Table Preference As
    This setting enables you to name the table preference. The preference setting is then selectable from PreferencesTable Preferences.
    PreferencesSave Table Preference
    This setting becomes the standard view for the user account. To restore default table columns, click PreferencesRestore Defaults.
The table preference remains until you apply another table preference or click Restore Defaults.